CAS RN 3877-15-4
CAS RN 3877-15-4
IUPAC Name:
1-(methylsulfanyl)propane
Synonyms:
N-propylmethylsulfide
Molecular Weight (g/mol):
90.18
Molecular Formula:
C4H10S
InChi Key:
ZOASGOXWEHUTKZ-UHFFFAOYSA-N
SMILES:
CCCSC
1
–
1
of
1
results
1
Methyl Propyl Sulfide, TRC
CAS: 3877-15-4 Molecular Formula: C4H10S Molecular Weight (g/mol): 90.18 IUPAC Name: 1-methylsulfanylpropane SMILES: CCCSC