CAS RN 387350-46-1
IUPAC Name:
3-(4-methanesulfonylphenyl)propanoic acid
Synonyms:
3-(4-methanesulfonylphenyl)propanoic acid
Molecular Weight (g/mol):
228.26
Molecular Formula:
C10H12O4S
InChi Key:
ZZZBVBATZOAPQB-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)C1=CC=C(CCC(O)=O)C=C1
1
–
1
of
1
results
1
3-[4-(Methylsulfonyl)phenyl]propionic acid, 96%, Thermo Scientific™
CAS: 387350-46-1 Molecular Formula: C10H12O4S Molecular Weight (g/mol): 228.26 MDL Number: MFCD01319236 InChI Key: ZZZBVBATZOAPQB-UHFFFAOYSA-N Synonym: 3-4-methylsulfonyl phenyl propanoic acid,3-4-methanesulfonylphenyl propanoic acid,3-4-methylsulphonylphenyl propionic acid,benzenepropanoic acid, 4-methylsulfonyl,4-methylsulfonyl benzenepropanoic acid,3-4-methylsulfonylphenyl propanoic acid,3-4-methylsulfonyl benzene-1-propanoic acid,3-4-methylsulfonyl phenyl propionic acid,3-4-methylsulphonyl phenyl propanoic acid PubChem CID: 2736960 IUPAC Name: 3-(4-methylsulfonylphenyl)propanoic acid SMILES: CS(=O)(=O)C1=CC=C(CCC(O)=O)C=C1