accessibility menu, dialog, popup

UserName

H3COON

CAS RN 38690-77-6

IUPAC Name: 4-cyanophenyl 4-butylbenzoate
Synonyms: 4-cyanophenyl 4-butylbenzoate
Molecular Weight (g/mol): 279.34
Molecular Formula: C18H17NO2
InChi Key: XQRFASOUJIKXRE-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C(=O)OC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results