accessibility menu, dialog, popup

UserName

CAS RN 386704-12-7

NOFFF

CAS RN 386704-12-7

IUPAC Name: 6-(trifluoromethyl)pyridine-3-carbaldehyde
Synonyms: 6-(trifluoromethyl)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 175.11
Molecular Formula: C7H4F3NO
InChi Key: MRPAGRCGPAXOGS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results