accessibility menu, dialog, popup

UserName

CAS RN 38663-85-3

CH3ONCS

CAS RN 38663-85-3

IUPAC Name: 1-isothiocyanato-2-methoxyethane
Synonyms: 1-isothiocyanato-2-methoxyethane
Molecular Weight (g/mol): 117.17
Molecular Formula: C4H7NOS
InChi Key: HTSAVXAFEVUJQE-UHFFFAOYSA-N
SMILES: COCCN=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results