accessibility menu, dialog, popup

UserName

CAS RN 38557-71-0

CH3NNCl

CAS RN 38557-71-0

IUPAC Name: 2-chloro-6-methylpyrazine
Synonyms: 2-chloro-6-methyl-pyrazine
Molecular Weight (g/mol): 128.56
Molecular Formula: C5H5ClN2
InChi Key: CKUVSPQGYLELRG-UHFFFAOYSA-N
SMILES: CC1=CN=CC(Cl)=N1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)