CAS RN 384342-65-8
CAS RN 384342-65-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Pyrenylmethyl Methanethiosulfonate, TRC
CAS: 384342-65-8 Molecular Formula: C18 H14 O2 S2 Molecular Weight (g/mol): 326.43 Synonym: Methanesulfonothioic acid, S-(1-pyrenylmethyl) ester,1-Pyrenylmethyl methanethiosulfonate IUPAC Name: 1-(methylsulfonylsulfanylmethyl)pyrene SMILES: CS(=O)(=O)SCc1ccc2ccc3cccc4ccc1c2c34