accessibility menu, dialog, popup

UserName

FFFOOH2C

CAS RN 383-67-5

IUPAC Name: prop-2-en-1-yl 2,2,2-trifluoroacetate
Synonyms: prop-2-en-1-yl 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 154.09
Molecular Formula: C5H5F3O2
InChi Key: XIVPVSIDXBTZLM-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)OCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results