accessibility menu, dialog, popup

UserName

CAS RN 381178-60-5

CH3NOOOOCl

CAS RN 381178-60-5

IUPAC Name: 5-(2-methyl-4-nitrophenyl)furan-2-carbonyl chloride
Synonyms: 5-(2-methyl-4-nitrophenyl)furan-2-carbonyl chloride
Molecular Weight (g/mol): 265.65
Molecular Formula: C12H8ClNO4
InChi Key: VBNSSVYGFDMJIZ-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(=C1)[N+]([O-])=O)C1=CC=C(O1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results