accessibility menu, dialog, popup

UserName

KOClOO

CAS RN 3811-04-9

IUPAC Name: potassium chlorate
Synonyms: potassium chlorate
Molecular Weight (g/mol): 122.55
Molecular Formula: ClKO3
InChi Key: VKJKEPKFPUWCAS-UHFFFAOYSA-M
SMILES: [K+].[O-][Cl](=O)=O
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
Percent Purity 
  • (9)
  • (2)
  • (2)
  • (2)
Grade 
  • (7)
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (5)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (5)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (9)
  • (2)
  • (2)
  • (2)
  • (7)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (4)