accessibility menu, dialog, popup

UserName

CAS RN 37972-69-3

IUPAC Name: 6-oxo-1,6-dihydropyridazine-3-carboxylate
Synonyms: 6-oxo-1H-pyridazine-3-carboxylate
Molecular Weight (g/mol): 139.09
Molecular Formula: C5H3N2O3
InChi Key: GIFSROMQVPUQFK-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=NNC(=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results