accessibility menu, dialog, popup

UserName

NaNH2(S)HOOOO

CAS RN 3792-50-5

IUPAC Name: sodium (2S)-2-amino-3-carboxypropanoate
Synonyms: sodium aspartate
Molecular Weight (g/mol): 155.09
Molecular Formula: C4H6NNaO4
InChi Key: WTWSHHITWMVLBX-DKWTVANSSA-M
SMILES: [Na+].N[C@@H](CC(O)=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)