accessibility menu, dialog, popup

UserName

CAS RN 37828-19-6

OO

CAS RN 37828-19-6

IUPAC Name: 1-phenylcyclobutane-1-carboxylate
Synonyms: 1-phenylcyclobutane-1-carboxylate
Molecular Weight (g/mol): 175.21
Molecular Formula: C11H11O2
InChi Key: JHZRNLRTNIDFKG-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1(CCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results