accessibility menu, dialog, popup

UserName

NNClFFF

CAS RN 37552-81-1

IUPAC Name: 4-chloro-6-(trifluoromethyl)pyrimidine
Synonyms: 4-chloro-6-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 182.53
Molecular Formula: C5H2ClF3N2
InChi Key: TYSPDLZOMUDHQZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results