accessibility menu, dialog, popup

UserName

CAS RN 375-50-8

FFIFFFFFFI

CAS RN 375-50-8

IUPAC Name: 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodobutane
Synonyms: 1,4-diiodo octafluorobutane
Molecular Weight (g/mol): 453.84
Molecular Formula: C4F8I2
InChi Key: JILAKKYYZPDQBE-UHFFFAOYSA-N
SMILES: FC(F)(I)C(F)(F)C(F)(F)C(F)(F)I
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)