CAS RN 3749-36-8
IUPAC Name:
(3,4-dihydro-2H-pyran-2-yl)methanol
Synonyms:
3,4-dihydro-2H-pyran-2-ylmethanol
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
XMICBFRKICBBKD-UHFFFAOYNA-N
SMILES:
OCC1CCC=CO1
1
–
1
of
1
results
1
3,4-Dihydro-2H-pyran-2-methanol, 96%
CAS: 3749-36-8 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.144 MDL Number: MFCD00051507 InChI Key: XMICBFRKICBBKD-UHFFFAOYSA-N Synonym: 3,4-dihydro-2h-pyran-2-methanol,2-hydroxymethyl-3,4-dihydro-2h-pyran,dhp linker,3,4-dihydro-2h-pyran-2-yl methanol,2h-pyran-2-methanol, 3,4-dihydro,dhp resin,ellmans dhp resin,zlchem 376,pubchem9703,acmc-209itf PubChem CID: 95559 IUPAC Name: 3,4-dihydro-2H-pyran-2-ylmethanol SMILES: C1CC(OC=C1)CO