accessibility menu, dialog, popup

UserName

CAS RN 37461-61-3

NNSBrFFF

CAS RN 37461-61-3

IUPAC Name: 2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole
Synonyms: 2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole
Molecular Weight (g/mol): 233.01
Molecular Formula: C3BrF3N2S
InChi Key: LGTVLLPQCMJOGT-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NN=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results