accessibility menu, dialog, popup

UserName

CAS RN 373603-19-1

IUPAC Name: 1-(2,4-difluoro-3-methoxyphenyl)ethan-1-one
Synonyms: 1-(2,4-difluoro-3-methoxyphenyl)ethanone
Molecular Weight (g/mol): 186.16
Molecular Formula: C9H8F2O2
InChi Key: KTZOZLGEZVDGMS-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(C(C)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results