accessibility menu, dialog, popup

UserName

CAS RN 372-29-2

CH3OOH2NFFF(Z)

CAS RN 372-29-2

IUPAC Name: ethyl (2Z)-3-amino-4,4,4-trifluorobut-2-enoate
Synonyms: ethyl (2Z)-3-amino-4,4,4-trifluorobut-2-enoate
Molecular Weight (g/mol): 183.13
Molecular Formula: C6H8F3NO2
InChi Key: NXVKRKUGIINGHD-ARJAWSKDSA-N
SMILES: CCOC(=O)\C=C(/N)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results