CAS RN 37159-60-7
CAS RN 37159-60-7
IUPAC Name:
3-bromo-5-chloro-1,2,4-thiadiazole
Synonyms:
3-bromo-5-chloro-1,2,4-thiadiazole
Molecular Weight (g/mol):
199.45
Molecular Formula:
C2BrClN2S
InChi Key:
CXUWGEWQRCXJDC-UHFFFAOYSA-N
SMILES:
ClC1=NC(Br)=NS1