accessibility menu, dialog, popup

UserName

CAS RN 37074-39-8

CH3OCH3Cl

CAS RN 37074-39-8

IUPAC Name: 1-(4-chloro-3-methylphenyl)ethan-1-one
Synonyms: 1-(4-chloro-3-methylphenyl)ethanone
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: XOIGZLJCLDWTQH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(C)=C(Cl)C=C1
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results