accessibility menu, dialog, popup

UserName

CH3OH3CFBFFFCH3

CAS RN 368-39-8

IUPAC Name: tetrafluoroboranuide; triethyloxidanium
Synonyms: triethyloxonium tetrafluoroborate
Molecular Weight (g/mol): 189.99
Molecular Formula: C6H15BF4O
InChi Key: IYDQMLLDOVRSJJ-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC[O+](CC)CC
Quantity 
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results