CAS RN 36761-83-8
CAS RN 36761-83-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Dechloroethyl Cyclophosphamide, TRC
CAS: 36761-83-8 Molecular Formula: C5 H12 Cl N2 O2 P Molecular Weight (g/mol): 198.59 Synonym: 2H-1,3,2-Oxazaphosphorin-2-amine, N-(2-chloroethyl)tetrahydro-, 2-oxide,2-(2-Chloroethylamino)tetrahydro-2H-1,3,2-oxazaphosphorine 2-oxide,3-Dechloroethylifosfamide,Asta 4968,Dechloroethylcyclophosphamide,Monochloroethylcyclophosphamide,N-Dechloroethylcyclophosphamide,(2RS)-2-[(2-Chloroethyl)amino]-1,3,2λ5-oxazaphosphinan-2-one IUPAC Name: N-(2-chloroethyl)-2-oxo-1,3,2λ^{5}-oxazaphosphinan-2-amine SMILES: ClCCNP1(=O)NCCCO1