accessibility menu, dialog, popup

UserName

CAS RN 36626-29-6

ClHOOP

CAS RN 36626-29-6

IUPAC Name: (2-carboxyethyl)triphenylphosphanium chloride
Synonyms: (2-carboxyethyl)triphenylphosphanium chloride
Molecular Weight (g/mol): 370.81
Molecular Formula: C21H20ClO2P
InChi Key: GALLWJZTZYJVSL-UHFFFAOYSA-N
SMILES: [Cl-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results