accessibility menu, dialog, popup

UserName

NCNNNa

CAS RN 36603-80-2

IUPAC Name: sodium tricyanomethanide
Synonyms: sodium tricyanomethanide
Molecular Weight (g/mol): 113.06
Molecular Formula: C4N3Na
InChi Key: LOYHZHALCDOXIK-UHFFFAOYSA-N
SMILES: [Na+].N#C[C-](C#N)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)