accessibility menu, dialog, popup

UserName

CH3NNH2N

CAS RN 365565-88-4

IUPAC Name: 4-{8-methylimidazo[1,2-a]pyridin-2-yl}aniline
Synonyms: 4-{8-methylimidazo[1,2-a]pyridin-2-yl}aniline
Molecular Weight (g/mol): 223.28
Molecular Formula: C14H13N3
InChi Key: GWBJQNMMOJBUIU-UHFFFAOYSA-N
SMILES: CC1=CC=CN2C=C(N=C12)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results