accessibility menu, dialog, popup

UserName

CAS RN 364794-85-4

BOOH3CCH3H3CH3CSNO

CAS RN 364794-85-4

IUPAC Name: 4-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl}morpholine
Synonyms: 4-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl}morpholine
Molecular Weight (g/mol): 309.23
Molecular Formula: C15H24BNO3S
InChi Key: AMUMGAXTCHBNPU-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CSC(CN2CCOCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results