CAS RN 3641-13-2
CAS RN 3641-13-2
IUPAC Name:
3-amino-1H-1,2,4-triazole-5-carboxylic acid
Synonyms:
5-amino-2H-1,2,4-triazole-3-carboxylic acid
Molecular Weight (g/mol):
128.09
Molecular Formula:
C3H4N4O2
InChi Key:
MVRGLMCHDCMPKD-UHFFFAOYSA-N
SMILES:
NC1=NNC(=N1)C(O)=O
1
–
3
of
3
results
1
5-Amino-4H-1,2,4-triazole-3-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Amino-4H-1,2,4-triazole-3-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Amino-4H-1,2,4-triazole-3-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.