accessibility menu, dialog, popup

UserName

HOOO

CAS RN 36330-85-5

IUPAC Name: 4-{[1,1'-biphenyl]-4-yl}-4-oxobutanoic acid
Synonyms: fenbufen cinopal wyeth brand of fenbufen fenbufen (product) lederfen fenbufenum 芬布芬 bufemid fenbufene fenbufen (substance)
Molecular Weight (g/mol): 254.29
Molecular Formula: C16H14O3
InChi Key: ZPAKPRAICRBAOD-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)