CAS RN 362602-54-8
IUPAC Name:
N-(2,6-dichlorophenyl)-2-methoxybenzamide
Synonyms:
N-(2,6-dichlorophenyl)-2-methoxybenzamide
Molecular Weight (g/mol):
296.15
Molecular Formula:
C14H11Cl2NO2
InChi Key:
FVOARQOPPNATSM-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1C(=O)NC1=C(Cl)C=CC=C1Cl
1
–
1
of
1
results
1
N-(2,6-Dichlorophenyl)-2-methoxybenzamide, 97%, Thermo Scientific™
CAS: 362602-54-8 Molecular Formula: C14H11Cl2NO2 Molecular Weight (g/mol): 296.147 MDL Number: MFCD01115478 InChI Key: FVOARQOPPNATSM-UHFFFAOYSA-N Synonym: n-2,6-dichlorophenyl-2-methoxybenzamide,n-2,6-dichlorophenyl 2-methoxyphenyl carboxamide PubChem CID: 4392873 IUPAC Name: N-(2,6-dichlorophenyl)-2-methoxybenzamide SMILES: COC1=CC=CC=C1C(=O)NC2=C(C=CC=C2Cl)Cl