accessibility menu, dialog, popup

UserName

CH3SBOHHO

CAS RN 362045-33-8

IUPAC Name: [2-(ethylsulfanyl)phenyl]boronic acid
Synonyms: 2-(ethylsulfanyl)phenylboronic acid
Molecular Weight (g/mol): 182.04
Molecular Formula: C8H11BO2S
InChi Key: SCGSNVLNGRYSAA-UHFFFAOYSA-N
SMILES: CCSC1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results