accessibility menu, dialog, popup

UserName

CAS RN 36140-83-7

CH3CH3NNON

CAS RN 36140-83-7

IUPAC Name: 3-(3,5-dimethyl-1H-pyrazol-1-yl)-3-oxopropanenitrile
Synonyms: 3-(3,5-dimethylpyrazol-1-yl)-3-oxopropanenitrile
Molecular Weight (g/mol): 163.18
Molecular Formula: C8H9N3O
InChi Key: DDWZYWSLHBDVGR-UHFFFAOYSA-N
SMILES: CC1=NN(C(=O)CC#N)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results