accessibility menu, dialog, popup

UserName

CAS RN 361394-74-3

CH3NHO

CAS RN 361394-74-3

IUPAC Name: methyl[(2-phenoxyphenyl)methyl]amine
Synonyms: methyl[(2-phenoxyphenyl)methyl]amine
Molecular Weight (g/mol): 213.28
Molecular Formula: C14H15NO
InChi Key: IMIUMEQDSKHKST-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1OC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results