accessibility menu, dialog, popup

UserName

CAS RN 36051-31-7

MgNH2NONN(R)O(R)OPOHOOPOHOOPHOOHO(S)OH(R)OHHN

CAS RN 36051-31-7

IUPAC Name: ({[({[(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid magnesium
Synonyms: guanosine triphosphate magnesium
Molecular Weight (g/mol): 547.49
Molecular Formula: C10H16MgN5O14P3
InChi Key: NMQPZZLDIUETGE-GWTDSMLYSA-N
SMILES: [Mg].NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O
Physical Form 
  • (10)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (5)
  • (5)
  • (10)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (6)