accessibility menu, dialog, popup

UserName

CAS RN 360-92-9

IUPAC Name: N,N-diethyl-2,2,2-trifluoroacetamide
Synonyms: N,N-diethyl-2,2,2-trifluoroacetamide
Molecular Weight (g/mol): 169.15
Molecular Formula: C6H10F3NO
InChi Key: CODXZFSZJFCVBE-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)