CAS RN 360-03-2
CAS RN 360-03-2
IUPAC Name:
2-(2,3-difluorophenyl)acetate
Synonyms:
2-(2,3-difluorophenyl)acetate
Molecular Weight (g/mol):
171.12
Molecular Formula:
C8H5F2O2
InChi Key:
UXSQXUSJGPVOKT-UHFFFAOYSA-M
SMILES:
[O-]C(=O)CC1=CC=CC(F)=C1F
1
–
2
of
2
results
1
alpha,alpha-Difluorophenylacetic acid, 97%
CAS: 360-03-2 Molecular Formula: C8H6F2O2 Molecular Weight (g/mol): 172.131 MDL Number: MFCD00498146 InChI Key: PFKSLFZFBCIJOI-UHFFFAOYSA-N PubChem CID: 726153 IUPAC Name: 2,2-difluoro-2-phenylacetic acid SMILES: C1=CC=C(C=C1)C(C(=O)O)(F)F
2
2,2-Difluoro-2-phenylacetic Acid, TRC
CAS: 360-03-2 Molecular Formula: C8H6F2O2 Molecular Weight (g/mol): 172.13 Synonym: alpha,alpha-Difluorobenzeneacetic Acid,Difluorophenylacetic Acid,alpha,alpha-Difluorophenylacetic Acid SMILES: OC(=O)C(F)(F)c1ccccc1