accessibility menu, dialog, popup

UserName

CAS RN 3586-15-0

IUPAC Name: 3-phenoxybenzoyl chloride
Synonyms: 3-phenoxybenzoyl chloride
Molecular Weight (g/mol): 232.66
Molecular Formula: C13H9ClO2
InChi Key: TTZXIWBOKOZOPL-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC(OC2=CC=CC=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results