accessibility menu, dialog, popup

UserName

CAS RN 35852-74-5

CH2NN

CAS RN 35852-74-5

IUPAC Name: 1-(prop-2-en-1-yl)-1H-pyrazole
Synonyms: PYRAZOLE1allyl
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: BPPPRRLOPGKVMQ-UHFFFAOYSA-N
SMILES: C=CCN1C=CC=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results