accessibility menu, dialog, popup

UserName

LiHH3CNHCH3

CAS RN 3585-33-9

IUPAC Name: lithium(1+) dimethylamine hydride
Synonyms: lithium(1+) dimethylamine hydride
Molecular Weight (g/mol): 53.03
Molecular Formula: C2H8LiN
InChi Key: QHXBFEDDNKOZHB-UHFFFAOYSA-N
SMILES: [H-].[Li+].CNC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)