accessibility menu, dialog, popup

UserName

FFFSOOOSOOFFF

CAS RN 358-23-6

IUPAC Name: trifluoromethanesulfonyl trifluoromethanesulfonate
Synonyms: triflic anhydride
Molecular Weight (g/mol): 282.13
Molecular Formula: C2F6O5S2
InChi Key: WJKHJLXJJJATHN-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F
Boiling Point 
  • (3)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (5)
  • (3)
  • (5)
  • (5)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)