accessibility menu, dialog, popup

UserName

CAS RN 357-57-3

IUPAC Name: (1R,18S,20R,21R)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one
Synonyms: brucine
Molecular Weight (g/mol): 394.47
Molecular Formula: C23H26N2O4
InChi Key: RRKTZKIUPZVBMF-PLNGPGDESA-N
SMILES: COC1=CC2=C(C=C1OC)[C@@]13CCN4CC5=CCOC6CC(=O)N2C1[C@H]6[C@H]5C[C@@H]34
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (9)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)