accessibility menu, dialog, popup

UserName

CH3OCl

CAS RN 35675-44-6

IUPAC Name: 2-(4-methylphenyl)acetyl chloride
Synonyms: 2-(4-methylphenyl)acetyl chloride
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: QDZAWVLWIMOXJT-UHFFFAOYSA-N
SMILES: CC1=CC=C(CC(Cl)=O)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)