accessibility menu, dialog, popup

UserName

CH3NHOOOHO

CAS RN 35661-39-3

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 311.34
Molecular Formula: C18H17NO4
InChi Key: QWXZOFZKSQXPDC-UHFFFAOYNA-N
SMILES: CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results