accessibility menu, dialog, popup

UserName

CAS RN 3564-73-6

IUPAC Name: 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-2-carboxamide
Synonyms: 10,11-dihydrocarbamazepine
Molecular Weight (g/mol): 238.29
Molecular Formula: C15H14N2O
InChi Key: PHNLCHMJDSSPDQ-UHFFFAOYSA-N
SMILES: NC(=O)N1C2=CC=CC=C2CCC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)