accessibility menu, dialog, popup

UserName

NaOHNHNOSOOO(Z)

CAS RN 3564-14-5

IUPAC Name: sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-1-sulfonate
Synonyms: sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-3-oxonaphthalene-1-sulfonate
Molecular Weight (g/mol): 416.38
Molecular Formula: C20H13N2NaO5S
InChi Key: YRFCCVCSEVISHI-GXTSIBQPSA-M
SMILES: [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O
Color 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)