accessibility menu, dialog, popup

UserName

CAS RN 3558-24-5

H3CN

CAS RN 3558-24-5

IUPAC Name: 1-methyl-2-phenyl-1H-indole
Synonyms: 1-methyl-2-phenylindole
Molecular Weight (g/mol): 207.28
Molecular Formula: C15H13N
InChi Key: SFWZZSXCWQTORH-UHFFFAOYSA-N
SMILES: CN1C(=CC2=CC=CC=C12)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results