accessibility menu, dialog, popup

UserName

BrOOBr

CAS RN 35578-47-3

IUPAC Name: bis(4-bromophenyl)ethane-1,2-dione
Synonyms: bis(4-bromophenyl)ethane-1,2-dione
Molecular Weight (g/mol): 368.02
Molecular Formula: C14H8Br2O2
InChi Key: NYCBYBDDECLFPE-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)