accessibility menu, dialog, popup

UserName

CAS RN 354-76-7

IUPAC Name: pentafluoropropanamide
Synonyms: pentafluoropropanamide
Molecular Weight (g/mol): 163.05
Molecular Formula: C3H2F5NO
InChi Key: KQTOYEUYHXUEDB-UHFFFAOYSA-N
SMILES: NC(=O)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results