accessibility menu, dialog, popup

UserName

CAS RN 354-65-4

IUPAC Name: 1,1,2,2-tetrafluoro-1,2-diiodoethane
Synonyms: 1,1,2,2-tetrafluoro-1,2-diiodoethane
Molecular Weight (g/mol): 353.83
Molecular Formula: C2F4I2
InChi Key: NZXVPCQHQVWOFD-UHFFFAOYSA-N
SMILES: FC(F)(I)C(F)(F)I
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results