accessibility menu, dialog, popup

UserName

CH3H3CCH3OCH3NH2

CAS RN 3535-88-4

IUPAC Name: 5-tert-butyl-2-methoxyaniline
Synonyms: 5-tert-butyl-2-methoxyaniline
Molecular Weight (g/mol): 179.26
Molecular Formula: C11H17NO
InChi Key: DLTLFDQLMFMTRQ-UHFFFAOYSA-N
SMILES: COC1=C(N)C=C(C=C1)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results